4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene ≥98.0% (by HPLC)
TCIAB4796-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):6.80 to 7.60 %
:
:581.76 g/mol
:883554-70-9
:C₄₂H₃₅N₃