1,3-Bis(4-bromophenyl)-2-propanone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:54523-47-6
:
Formula:C₁₅H₁₂Br₂O
MDL Number:MFCD03843365
:368.07 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:117.0 to 121.0 deg-C
Specification Test Results
Frequently Bought Together