
4-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
: TCI
:850568-54-6
:
:C₁₁H₁₅BO₄
:MFCD03411946
:222.05 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 109.0 %
Frequently Bought Together