Order Entry
Netherlands
ContactUsLinkComponent
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Irritant
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Catalog # TCIAB4770-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Catalog # TCIAB4770-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    105.0 to 109.0 deg-C

Specification Test Results

About this item

 :
 :229.00 g/mol
 :GHS07
CAS Number:60061-68-9
Formula:C₅H₄BrF₃N₂
MDL Number:MFCD08060951