4,7-Bis(5-bromo-4-n-octyl-2-thienyl)-2,1,3-benzothiadiazole ≥97.0% (by HPLC)
Catalog # TCIAB4684-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):3.90 to 4.40 %
Specification Test Results
About this item
:
:682.64 g/mol
CAS Number:457931-23-6
Formula:C₃₀H₃₈Br₂N₂S₃