Order Entry
Netherlands
ContactUsLinkComponent
4,4'-Bis[4-(di-p-tolylamino)styryl]biphenyl ≥98.0% (by HPLC)
Irritant
4,4'-Bis[4-(di-p-tolylamino)styryl]biphenyl ≥98.0% (by HPLC)
Catalog # TCIAB4682-200MG
Supplier:  TCI
CAS Number:  
Irritant
4,4'-Bis[4-(di-p-tolylamino)styryl]biphenyl ≥98.0% (by HPLC)
Catalog # TCIAB4682-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

About this item

 :
 :749.01 g/mol
 :GHS07
CAS Number:119586-44-6
Formula:C₅₆H₄₈N₂
MDL Number:MFCD09751239