1,4-Bis(2-propynyloxy)benzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34596-36-6
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00224313
:186.21 g/mol
Boiling Pt:202°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:48.0 to 52.0 deg-C
Specification Test Results
Frequently Bought Together