Order Entry
Netherlands
ContactUsLinkComponent
1,3-Bis[(di-tert-butylphosphino)oxy]benzene ≥94.0% (by GC)
Irritant
1,3-Bis[(di-tert-butylphosphino)oxy]benzene ≥94.0% (by GC)
Catalog # TCIAB4595-1G
Supplier:  TCI
CAS Number:  
Irritant
1,3-Bis[(di-tert-butylphosphino)oxy]benzene ≥94.0% (by GC)
Catalog # TCIAB4595-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 94.0 %

Specification Test Results

About this item

 :
 :398.51 g/mol
 :GHS07
CAS Number:338800-20-7
Formula:C₂₂H₄₀O₂P₂