2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene ≥98.0% (by HPLC)
Catalog # TCIAB4588-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:116.0 to 120.0 deg-C
Specification Test Results
About this item
:
:698.69 g/mol
CAS Number:711026-06-1
Formula:C₄₅H₇₂B₂O₄
MDL Number:MFCD20275090