Order Entry
Netherlands
ContactUsLinkComponent
9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together