2-(3-Bromophenyl)naphthalene ≥95.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:667940-23-0
:
Formule:C₁₆H₁₁Br
MDL-nummer:MFCD16658911
:283.168 g/mol
Specificatie testresultaten
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:97.0 to 101.0 deg-C
Specificatie testresultaten
Frequently Bought Together