Order Entry
Netherlands
ContactUsLinkComponent
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne ≥98.0% (by GC)
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne ≥98.0% (by GC)
Catalog # TCIAB4561-1G
Supplier:  TCI
CAS Number:  
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne ≥98.0% (by GC)
Catalog # TCIAB4561-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :277.96 g/mol
CAS Number:1010840-17-1
Formula:C₁₄H₂₄B₂O₄
MDL Number:MFCD22666429