
3-Bromo-5-(trifluoromethyl)benzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:328-67-6
:
Formula:C₈H₄BrF₃O₂
MDL Number:MFCD03412186
:269.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
Specification Test Results
Frequently Bought Together