Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,8-Bis(diphenylphosphino)naphthalene ≥97.0% (by GC)
 :  TCI
 
 
1,8-Bis(diphenylphosphino)naphthalene ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together