2-Bromophenyl p-Toluenesulphonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:84672-48-0
:
Formula:C₁₃H₁₁BrO₃S
MDL Number:MFCD19707445
:327.19 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:74.0 to 78.0 deg-C
Specification Test Results
Frequently Bought Together