1,3-Bis(2-propynyloxy)benzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:26627-36-1
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00224312
:186.21 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:38.0 to 42.0 deg-C
Specification Test Results
Frequently Bought Together