Order Entry
Netherlands
ContactUsLinkComponent
2,2'-Bicinchoninic acid disodium salt, hydrate ≥98.0% (by HPLC, titration analysis)
Irritant
2,2'-Bicinchoninic acid disodium salt, hydrate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2,2'-Bicinchoninic acid disodium salt, hydrate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    3.0 to 16.0 %

Specification Test Results

Frequently Bought Together