
[[(4-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥97.0% (by HPLC)
Catalog # TCIAB4489-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):7.00 to 8.00 %
- Melting point:184.0 to 188.0 deg-C
- Lambda max.:514.0 to 519.0 nm (CH3CN)
Specification Test Results
About this item
:
:374.33 g/mol
:GHS07
CAS Number:151486-56-5
Formula:C₂₂H₃₃BF₂N₂