

2,6-Bis(trimethylstannyl)-4,8-bis[(2-n-octyldodecyl)oxy]benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
Catalog # TCIAB4488-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light gray to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
About this item
:
:1108.97 g/mol
UN:3146
ADR:6.1,I
:GHS06
:GHS09
CAS Number:1320201-22-6
Formula:C₅₆H₁₀₂O₂S₂Sn₂