1,3-Bis(4-methoxyphenyl)thiourea ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB4482-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:185.0 to 189.0 deg-C
Specification Test Results
About this item
:
:288.37 g/mol
CAS Number:1227-45-8
Formula:C₁₅H₁₆N₂O₂S
MDL Number:MFCD00025792