Order Entry
Netherlands
ContactUsLinkComponent
9-(4-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)
9-(4-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)
Supplier:  TCI
Certificates
9-(4-Bromophenyl)-10-phenylanthracene ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    225.0 to 229.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together