Order Entry
Netherlands
ContactUsLinkComponent
4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole ≥95.0% (by GC)
Toxic
Environment
4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole ≥95.0% (by GC)
Catalog # TCIAB4453-200MG
Supplier:  TCI
CAS Number:  
Toxic
Environment
4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole ≥95.0% (by GC)
Catalog # TCIAB4453-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :626.03 g/mol
UN:3146
ADR:6.1,I
 :GHS06
 :GHS09
CAS Number:1025451-57-3
Formula:C₂₀H₂₄N₂S₃Sn₂
MDL Number:MFCD28049209