9-Bromo-10-(naphthalen-1-yl)anthracene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:400607-04-7
:
Formula:C₂₄H₁₅Br
MDL Number:MFCD11046571
:383.29 g/mol
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together