Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl (piperidin-4-ylmethyl)carbamate ≥98.0% (by GC, titration analysis)
Corrosive
tert-Butyl (piperidin-4-ylmethyl)carbamate ≥98.0% (by GC, titration analysis)
Catalog # TCIAB4444-1G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
tert-Butyl (piperidin-4-ylmethyl)carbamate ≥98.0% (by GC, titration analysis)
Catalog # TCIAB4444-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    107.0 to 111.0 deg-C

Specification Test Results

About this item

 :
 :214.31 g/mol
UN:3259
ADR:8,III
 :GHS05
CAS Number:135632-53-0
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD01631214