
1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:94525-05-0
:
Formula:C₂₀H₁₄F₆N₂O₂
MDL Number:MFCD07368071
:428.33 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:132.0 to 136.0 deg-C
Specification Test Results
Frequently Bought Together