
4,4'-Bis(5-hexyl-2-thienyl)-2,2'-bipyridyl ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB4420-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:488.75 g/mol
:GHS07
CAS Number:1047684-56-9
Formula:C₃₀H₃₆N₂S₂
MDL Number:MFCD22581259