Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
 
 
Boron trichloride ∼17% ∼1 mol/L in hexane
  TCIAB4416-100ML
 :  TCI
 :  
 
 
 
 
 
Boron trichloride ∼17% ∼1 mol/L in hexane
  TCIAB4416-100ML
 :  TCI
 :  

 

  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Neutralization titration):
    16.0 to 17.5 w/w%

 

 

 :
 :117.17 g/mol
 :14,01348
 :Refrigerator
 :233-658-4
 :2924
 :3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :GHS09
 :10294-34-5
 :BCl₃
 :MFCD00011313