Order Entry
Netherlands
ContactUsLinkComponent
1-Butene ∼10% in hexane
Flammable
Irritant
Health
1-Butene ∼10% in hexane
Supplier:  TCI
Certificates
Flammable
Irritant
Health
1-Butene ∼10% in hexane
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (HNMR):
    10.0 to 12.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together