4,8-Bis-n-octyloxybenzo[1,2-b:4,5-b']dithiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1098102-94-3
:
Formula:C₂₆H₃₈O₂S₂
MDL Number:MFCD26793498
:446.71 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:46.0 to 50.0 deg-C
Specification Test Results
Frequently Bought Together