


1,3-Butadiene ∼13% in hexane
Supplier: TCI
Certificates
About this item
CAS Number:106-99-0
:
Formula:C₄H₆
MDL Number:MFCD00008659
:54.09 g/mol
Merck Index:14,01509
Storage Temperature:Refrigerator
EINECS:203-450-8
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (HNMR):15.0 to 18.0 w/w%
Specification Test Results
Frequently Bought Together