
5-Bromo-2-iodobenzoic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAB4296-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:160.0 to 164.0 deg-C
Specification Test Results
About this item
:
:326.92 g/mol
:GHS07
CAS Number:21740-00-1
Formula:C₇H₄BrIO₂
MDL Number:MFCD00079717