Order Entry
Netherlands
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid ≥98.0% (by GC, titration analysis)
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    55.0 to 59.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together