N-(tert-Butoxycarbonyl)-6-aminohexanoic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6404-29-1
:
Formula:C₁₁H₂₁NO₄
MDL Number:MFCD00037798
:231.29 g/mol
Boiling Pt:166°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together