Order Entry
Netherlands
ContactUsLinkComponent
1,1'-Bis(dichlorophosphino)ferrocene ≥95.0% (by titrimetric analysis)
 
1,1'-Bis(dichlorophosphino)ferrocene ≥95.0% (by titrimetric analysis)
  TCIAB4272-200MG
 :  TCI
 :  
 
1,1'-Bis(dichlorophosphino)ferrocene ≥95.0% (by titrimetric analysis)
  TCIAB4272-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (Argentmetric Titration):
    min. 95.0 %
  • Purity (Chelometric Titration):
    min. 95.0 %
  • Melting point:
    68.0 to 72.0 deg-C

 

 

 :
 :8,II
 :387.77 g/mol
 :3261
 :GHS05
 :142691-70-1
 :C₁₀H₈Cl₄FeP₂
 :MFCD01630854