Order Entry
Netherlands
ContactUsLinkComponent
Dithiodiglycol (bis(2-hydroxyethyl) disulphide) ≥98.0% (by GC)
Toxic
Dithiodiglycol (bis(2-hydroxyethyl) disulphide) ≥98.0% (by GC)
Catalog # TCIAB4263-25G
Supplier:  TCI
CAS Number:  
Toxic
Dithiodiglycol (bis(2-hydroxyethyl) disulphide) ≥98.0% (by GC)
Catalog # TCIAB4263-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid to cloudy liquid
  • Purity (GC):
    min. 98.0 %
  • Water:
    max. 1.0 %

Specification Test Results

About this item

 :
 :154.25 g/mol
Boiling Pt:163°C
Flash Pt:112°C
EINECS:217-576-6
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:1892-29-1
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00002906