
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:114214-69-6
:
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD02179040
:201.27 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together