Boc-N-Me-Val-OH ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB4248-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-85.0 to -91.0 deg (C=0.5, EtOH)
Specification Test Results
About this item
:
:231.292 g/mol
CAS Number:45170-31-8
Formula:C₁₁H₂₁NO₄
MDL Number:MFCD00038760