Order Entry
Netherlands
ContactUsLinkComponent
N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine ≥98.0% (by GC, titration analysis)
Irritant
N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    100.0 to 104.0 deg-C

Specification Test Results

Frequently Bought Together