
(R)-1,1'-Binaphthyl-2,2'-disulphonyl dichloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1187629-43-1
:
Formula:C₂₀H₁₂Cl₂O₄S₂
:451.33 g/mol
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 99.0 ee%
- Specific rotation [a]20/D:-42.0 to -46.0 deg (C=1、THF)
Specification Test Results
Frequently Bought Together