
1,4-Benzodioxane-6-boronic acid (contains varying amounts of Anhydride)
Catalog # TCIAB4181-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
About this item
:
:179.97 g/mol
Melting Pt:118°C
:GHS07
CAS Number:164014-95-3
Formula:C₈H₉BO₄
MDL Number:MFCD01009696