Order Entry
Netherlands
ContactUsLinkComponent
cis-1,3-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)
Corrosive
Irritant
cis-1,3-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Corrosive
Irritant
cis-1,3-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together