Order Entry
Netherlands
ContactUsLinkComponent
Fmoc-Dap(Boc)-OH ≥97.0% (by HPLC, titration analysis)
Fmoc-Dap(Boc)-OH ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAB4174-1G
Supplier:  TCI
CAS Number:  
Fmoc-Dap(Boc)-OH ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAB4174-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 % (calcd.on anh.substance)
  • Specific rotation [a]20/D:
    -7.0 to -9.0 deg (C=, MeOH) (calcd.on anh.substance)
  • Water:
    3.0 to 6.0 %

Specification Test Results

About this item

 :
 :426.47 g/mol
Melting Pt:63°C
Storage Temperature:Refrigerator
CAS Number:162558-25-0
Formula:C₂₃H₂₆N₂O₆
MDL Number:MFCD00235896