
β-Methasone sodium phosphate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB4110-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:+99.0 to +105.0 deg (C=1, H2O (calcd.on anh.substance))
- Water:max. 10.0 %
Specification Test Results
About this item
:
:516.41 g/mol
Merck Index:14,01180
EINECS:205-797-0
:GHS07
CAS Number:151-73-5
Formula:C₂₂H₂₈FO₈P·₂Na
MDL Number:MFCD00200361