Order Entry
Netherlands
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -33.0 to -36.0 deg (C=1,CHCl3)

Specification Test Results

Frequently Bought Together