
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:71026-66-9
:
Formula:C₁₁H₁₆N₂O₂
MDL Number:MFCD00043022
:208.26 g/mol
EINECS:275-132-7
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:114.0 to 118.0 deg-C
Specification Test Results
Frequently Bought Together