(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:93267-04-0
:
Formula:C₉H₁₆INO₄
MDL Number:MFCD00216579
:329.13 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:49.0 to 53.0 deg-C
- Specific rotation [a]20/D:-3.0 to -5.0 deg (C=3、MeOH)
Specification Test Results
Frequently Bought Together