tert-Butyl-3-oxocyclobutylcarbamate ≥98.0% (by GC)
TCIAB4043-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:121.0 to 125.0 deg-C
:
:185.22 g/mol
:154748-49-9
:C₉H₁₅NO₃
:MFCD09751872