
N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB4028-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:99.0 to 103.0 deg-C
Specification Test Results
About this item
:
:210.24 g/mol
:GHS07
CAS Number:152120-61-1
Formula:C₉H₁₄N₄O₂
MDL Number:MFCD00216663