Order Entry
Netherlands
ContactUsLinkComponent
1-Boc-4-(methylamino)piperidine ≥98.0% (by GC, titration analysis)
Irritant
1-Boc-4-(methylamino)piperidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
1-Boc-4-(methylamino)piperidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together