Order Entry
Netherlands
ContactUsLinkComponent
4-Butylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Corrosive
4-Butylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Corrosive
4-Butylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together